siRNA Details
| Gene Symbol: ADGRG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308362.1 | NP_001295291.1 | 10 | 2172 | ||
| NM_032787.2 | NP_116176.2 | 16 | 2996 | |||
| XM_011513245.2 | XP_011511547.1 | 15 | 2871 | |||
| GenBank mRNA | AK027360.1 | - | 16 | 2984 | ||
| AK131069.1 | - | 9 | 1855 | |||
| AK172769.1 | - | 12 | 2238 | |||
| AK172774.1 | - | 8 | 1749 | |||
More Information
Set Membership: |
|

ADGRG7
Genomic Map