siRNA Details
| Gene Symbol: Prl7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164058.1 | NP_001157530.1 | 2, 3 | 254 | ||
| NM_008930.2 | NP_032956.1 | 2, 3 | 251 | |||
| GenBank mRNA | AF011381.1 | - | 2, 3 | 243 | ||
| AF020525.1 | - | 2, 3 | 225 | |||
| AK028371.1 | - | 2, 3 | 252 | |||
| AK141309.1 | - | 2, 3 | 255 | |||
| AK146050.1 | - | 2, 3 | 255 | |||
| BB611867.1 | - | 2, 3 | 252 | |||
| BC051671.1 | - | 2, 3 | 236 | 30018275 | ||

Prl7a1
Genomic Map