siRNA Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008944.2 | NP_032970.2 | 5 | 464 | ||
| GenBank mRNA | AF099733.1 | - | 5 | 431 | ||
| AK012119.1 | - | 5 | 446 | |||
| AK012990.1 | - | 5 | 453 | |||
| AK035578.1 | - | 453 | ||||
| AK136115.1 | - | 5 | 466 | |||
| AK151883.1 | - | 5 | 455 | |||
| AK152862.1 | - | 5 | 455 | |||
| AK154011.1 | - | 5 | 448 | |||
| AK167752.1 | - | 5 | 455 | |||
| BC026768.1 | - | 5 | 437 | 3156907 | ||
| CJ052982.1 | - | 5 | 466 | |||
| X70303.1 | - | 5 | 508 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_017279, J02897, BC060576, FQ214776, FQ223872.

Psma2
Genomic Map