siRNA Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008960.2 | NP_032986.1 | 2, 3 | 1040 | ||
| GenBank mRNA | AK076980.1 | - | 2, 3 | 998 | ||
| AK088717.1 | - | 2, 3 | 1019 | |||
| AK148736.1 | - | 2, 3 | 875 | |||
| AK164302.1 | - | 2, 3 | 1023 | |||
| AK208703.1 | - | 2, 3 | 272 | |||
| BC021445.1 | - | 2, 3 | 389 | 5038272 | ||
| U92437.1 | - | 2, 3 | 1120 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031606, AF017185, AF455569.

Pten
Genomic Map