siRNA Details
| Gene Symbol: Ptpn20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008978.2 | NP_033004.1 | 12 | 2639 | ||
| GenBank mRNA | AK029493.1 | - | 12 | 2638 | ||
| D64141.1 | - | 11 | 2639 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn20| Gene Symbol: Ptpn20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008978.2 | NP_033004.1 | 12 | 2639 | ||
| GenBank mRNA | AK029493.1 | - | 12 | 2638 | ||
| D64141.1 | - | 11 | 2639 | |||