siRNA Details
| Gene Symbol: Ptrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008986.2 | NP_033012.1 | 2 | 879 | ||
| GenBank mRNA | AF036249.1 | - | 2 | 834 | ||
| BC110698.1 | - | 2 | 861 | 30608521 | ||
| BC116701.1 | - | 2 | 821 | 40104877 | ||
| BC117527.1 | - | 2 | 821 | 40104876 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001105841.

Ptrf
Genomic Map