siRNA Details
| Gene Symbol: Rbpjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009036.1 | NP_033062.1 | 3 | 330 | ||
| GenBank mRNA | AK052449.1 | - | 3 | 1456 | ||
| AK131814.1 | - | 3 | 311 | |||
| AK131832.1 | - | 3 | 316 | |||
| BC152311.1 | - | 3 | 324 | 40090714 | ||
| Y10926.1 | - | 3 | 330 | |||

Rbpjl
Genomic Map