siRNA Details
| Gene Symbol: Rdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009040.3 | NP_033066.1 | 4 | 1406 | ||
| GenBank mRNA | AF030513.1 | - | 4 | 1406 | ||
| AK149404.1 | - | 3 | 1150 | |||
| AK164728.1 | - | 1 | 585 | |||
| BC089612.1 | - | 4 | 1375 | 6763439 | ||

Rdh16
Genomic Map