siRNA Details
| Gene Symbol: Rit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163310.1 | NP_001156782.1 | 3, 4 | 238 | ||
| NM_009069.4 | NP_033095.1 | 4, 5 | 383 | |||
| GenBank mRNA | AK005357.1 | - | 4, 5 | 344 | ||
| AK014126.1 | - | 4, 5 | 374 | |||
| AK164196.1 | - | 4, 5 | 374 | |||
| AK184013.1 | - | 1, 2 | 38 | |||
| BC012694.1 | - | 4, 5 | 343 | 4023732 | ||
| BY284832.1 | - | 4, 5 | 386 | |||
| CV562852.1 | - | 4, 5 | 351 | |||
| CX242151.1 | - | 3, 4 | 186 | |||
| U71205.1 | - | 4, 5 | 320 | |||

Rit1
Genomic Map