siRNA Details
| Gene Symbol: Rbbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009031.3 | NP_033057.3 | 3 | 502 | ||
| GenBank mRNA | AK076016.1 | - | 3 | 427 | ||
| AK078349.1 | - | 3 | 415 | |||
| AK082892.1 | - | 3 | 415 | |||
| AK132101.1 | - | 3 | 412 | |||
| AK135779.1 | - | 3 | 416 | |||
| AK135869.1 | - | 2 | 111 | |||
| AK135875.1 | - | 2 | 111 | |||
| AK135956.1 | - | 3 | 416 | |||
| AK136110.1 | - | 3 | 414 | |||
| AK145531.1 | - | 3 | 413 | |||
| AK145651.1 | - | 3 | 396 | |||
| AK146014.1 | - | 3 | 414 | |||
| AK146439.1 | - | 3 | 473 | |||
| AK146904.1 | - | 3 | 415 | |||
| AK146967.1 | - | 3 | 414 | |||
| AK147062.1 | - | 3 | 473 | |||
| AK148852.1 | - | 3 | 442 | |||
| AK153913.1 | - | 3 | 459 | |||
| AK160023.1 | - | 3 | 414 | |||
| BC003785.1 | - | 3 | 431 | 3600013 | ||
| U35142.1 | - | 3 | 441 | |||

Rbbp7
Genomic Map