siRNA Details
| Gene Symbol: Rex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177767.1 | NP_001171238.1 | 5 | 578 | ||
| GenBank mRNA | AF051348.1 | - | 573 | |||
| BC067397.1 | - | 3 | 333 | 30462971 | ||
| Gene Symbol: Gm13242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103158.2 | NP_001096628.2 | 5 | 578 | ||
| GenBank mRNA | BC096593.1 | - | 5 | 590 | 30459933 | |

Rex2
Genomic Map