siRNA Details
| Gene Symbol: Rit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163310.1 | NP_001156782.1 | 4 | 289 | ||
| NM_009069.4 | NP_033095.1 | 5 | 434 | |||
| GenBank mRNA | AK005357.1 | - | 5 | 395 | ||
| AK014126.1 | - | 5 | 425 | |||
| AK018785.1 | - | 4 | 593 | |||
| AK164196.1 | - | 5 | 425 | |||
| AK168254.1 | - | 5 | 504 | |||
| BC012694.1 | - | 5 | 394 | 4023732 | ||
| CV562852.1 | - | 5 | 402 | |||
| CX242151.1 | - | 4 | 237 | |||
| U71205.1 | - | 5 | 371 | |||

Rit1
Genomic Map