siRNA Details
| Gene Symbol: Rnps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080127.1 | NP_001073596.1 | 8 | 1775 | ||
| NM_001080128.1 | NP_001073597.1 | 7 | 1594 | |||
| NM_009070.2 | NP_033096.2 | 9 | 1897 | |||
| GenBank mRNA | AK011475.1 | - | 8 | 1771 | ||
| AK146443.1 | - | 7 | 1593 | |||
| AK151150.1 | - | 8 | 1765 | |||
| AK165905.1 | - | 7 | 1581 | |||
| BC002061.1 | - | 8 | 1763 | 3485311 | ||
| BC008089.1 | - | 5 | 1332 | |||
| X70067.1 | - | 8 | 1785 | |||

Rnps1
Genomic Map