siRNA Details
| Gene Symbol: Frrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113478.1 | NP_001106950.1 | 4 | 394 | ||
| NM_009146.3 | NP_033172.2 | 4 | 356 | |||
| GenBank mRNA | AK075573.1 | - | 4 | 348 | ||
| AK144933.1 | - | 4 | 348 | |||
| AK162693.1 | - | 4 | 349 | |||
| AK167289.1 | - | 4 | 384 | |||
| AK213195.1 | - | 2 | 47 | |||
| AU067508.1 | - | 4 | 356 | |||
| BC027770.1 | - | 3 | 354 | 5344922 | ||
| D50464.1 | - | 4 | 344 | |||

Frrs1
Genomic Map