siRNA Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009180.3 | NP_033206.2 | 3 | 404 | ||
| GenBank mRNA | AK004613.1 | - | 3 | 405 | ||
| AK136381.1 | - | 407 | ||||
| BC003276.1 | - | 3 | 1306 | |||
| BC010208.1 | - | 356 | ||||
| X93999.1 | - | 3 | 368 | |||

St6galnac2
Genomic Map