siRNA Details
Gene Symbol: Siah1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009172.2 | NP_033198.1 | 2 | 1766 | ||
GenBank mRNA | AK019266.1 | - | 1 | 564 | ||
Z19579.1 | - | 2 | 1694 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Siah1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009172.2 | NP_033198.1 | 2 | 1766 | ||
GenBank mRNA | AK019266.1 | - | 1 | 564 | ||
Z19579.1 | - | 2 | 1694 |