siRNA Details
| Gene Symbol: Rsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009105.4 | NP_033131.2 | 9 | 1298 | ||
| GenBank mRNA | AK009288.1 | - | 9 | 1188 | ||
| AK013076.1 | - | 5 | 769 | |||
| AK014749.1 | - | 4 | 641 | |||
| AK149835.1 | - | 9 | 1171 | |||
| AK150011.1 | - | 9 | 1166 | |||
| AK152655.1 | - | 9 | 1178 | |||
| BC003827.1 | - | 9 | 1161 | 3493122 | ||
| X63039.1 | - | 9 | 1112 | |||

Rsu1
Genomic Map