siRNA Details
| Gene Symbol: Sap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009119.3 | NP_033145.2 | 4 | 3251 | ||
| GenBank mRNA | Z97062.1 | - | 4 | 3186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sap18| Gene Symbol: Sap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009119.3 | NP_033145.2 | 4 | 3251 | ||
| GenBank mRNA | Z97062.1 | - | 4 | 3186 | ||