siRNA Details
| Gene Symbol: Satb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163630.1 | NP_001157102.1 | 5 | 911 | ||
| NM_001163631.1 | NP_001157103.1 | 5 | 779 | |||
| NM_001163632.1 | NP_001157104.1 | 5 | 752 | |||
| NM_009122.2 | NP_033148.2 | 5 | 802 | |||
| GenBank mRNA | AK037740.1 | - | 5 | 802 | ||
| AK088563.1 | - | 5 | 778 | |||
| AK134040.1 | - | 5 | 782 | |||
| AK158518.1 | - | 5 | 912 | |||
| BC011132.1 | - | 5 | 662 | 4164993 | ||
| CT010371.1 | - | 4 | 590 | |||
| U05252.1 | - | 5 | 788 | |||

Satb1
Genomic Map