siRNA Details
| Gene Symbol: Scg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164790.1 | NP_001158262.1 | 3 | 498 | ||
| NM_009130.3 | NP_033156.1 | 3 | 498 | |||
| GenBank mRNA | AK133655.1 | - | 3 | 400 | ||
| AK139140.1 | - | 3 | 438 | |||
| AK148498.1 | - | 3 | 480 | |||
| AK158315.1 | - | 3 | 371 | |||
| AK164089.1 | - | 3 | 441 | |||
| BC024785.1 | - | 3 | 457 | 5362975 | ||
| BU058741.1 | - | 3 | 498 | |||
| U02982.1 | - | 3 | 462 | |||

Scg3
Genomic Map