siRNA Details
| Gene Symbol: Slc16a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009197.2 | NP_033223.2 | 6 | 3931 | ||
| GenBank mRNA | AF045692.1 | - | 6 | 3998 | ||
| AK143100.1 | - | 1 | 1298 | |||
| BC080678.1 | - | 6 | 3767 | 6313495 | ||
| BC096587.1 | - | 6 | 3884 | 3992886 | ||

Slc16a2
Genomic Map