siRNA Details
| Gene Symbol: Slc22a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009202.5 | NP_033228.2 | 5 | 1089 | ||
| GenBank mRNA | AF010259.1 | - | 5 | 947 | ||
| BC021651.1 | - | 5 | 1089 | 4211979 | ||
| U38652.1 | - | 5 | 944 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a1| Gene Symbol: Slc22a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009202.5 | NP_033228.2 | 5 | 1089 | ||
| GenBank mRNA | AF010259.1 | - | 5 | 947 | ||
| BC021651.1 | - | 5 | 1089 | 4211979 | ||
| U38652.1 | - | 5 | 944 | |||