siRNA Details
| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009270.3 | NP_033296.1 | 2 | 1199 | ||
| GenBank mRNA | AK138457.1 | - | 2 | 1210 | ||
| AK148419.1 | - | 2 | 1215 | |||
| AK161837.1 | - | 2 | 1185 | |||
| AK163502.1 | - | 2 | 1179 | |||
| AK179451.1 | - | 1 | 82 | |||
| AK181007.1 | - | 1 | 82 | |||
| BC042781.1 | - | 2 | 456 | 4915911 | ||
| BC056361.1 | - | 2 | 1199 | 6816800 | ||
| D42048.1 | - | 2 | 1065 | |||

Sqle
Genomic Map