siRNA Details
| Gene Symbol: GPR160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014373.2 | NP_055188.1 | 4 | 837 | ||
| XM_005247346.3 | XP_005247403.1 | 5 | 1447 | |||
| XM_005247347.3 | XP_005247404.1 | 3 | 483 | |||
| XM_006713582.3 | XP_006713645.1 | 4 | 799 | |||
| XM_011512678.2 | XP_011510980.1 | 5 | 1129 | |||
| XM_017006161.1 | XP_016861650.1 | 5 | 940 | |||
| XM_017006162.1 | XP_016861651.1 | 5 | 944 | |||
| XM_017006163.1 | XP_016861652.1 | 4 | 1461 | |||
| GenBank mRNA | AJ249248.1 | - | 3 | 568 | ||
| BC000181.2 | - | 4 | 772 | 3048193 | ||

GPR160
Genomic Map