siRNA Details
| Gene Symbol: NDUFA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015965.6 | NP_057049.5 | 1 | 52 | ||
| GenBank mRNA | AF261134.1 | - | 1 | 483 | ||
| AF286697.1 | - | 1 | 30 | |||
| AK293859.1 | - | 1 | 34 | |||
| BC000589.2 | - | 1 | 31 | |||
| BC009189.2 | - | 1 | 39 | 3957082 | ||
| BG705222.1 | - | 1 | 41 | |||
| BG724220.1 | - | 1 | 527 | |||
| BM839920.1 | - | 1 | 52 | |||
| BP230223.1 | - | 1 | 32 | |||

NDUFA13
Genomic Map