siRNA Details
| Gene Symbol: Sos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009231.2 | NP_033257.2 | 3 | 828 | ||
| GenBank mRNA | AK140310.1 | - | 3 | 828 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sos1| Gene Symbol: Sos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009231.2 | NP_033257.2 | 3 | 828 | ||
| GenBank mRNA | AK140310.1 | - | 3 | 828 | ||