siRNA Details
| Gene Symbol: CRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005206.4 | NP_005197.3 | 2 | 527 | ||
| NM_016823.3 | NP_058431.2 | 2 | 527 | |||
| GenBank mRNA | AK291060.1 | - | 2 | 492 | ||
| BC001718.1 | - | 2 | 479 | 3353237 | ||
| BC008506.1 | - | 2 | 510 | 3452137 | ||
| BC009837.1 | - | 2 | 492 | 4040536 | ||
| BT007277.1 | - | 2 | 377 | |||
| D10656.1 | - | 2 | 482 | |||
| KU177996.1 | - | 2 | 377 | |||
| KU177997.1 | - | 2 | 377 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001277219, NM_133656, BC012216, AK152549, AK052795, and others.

CRK
Genomic Map