siRNA Details
| Gene Symbol: Slk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164639.1 | NP_001158111.1 | 2 | 372 | ||
| NM_009289.3 | NP_033315.2 | 2 | 372 | |||
| GenBank mRNA | AF039574.1 | - | 2 | 377 | ||
| AF112855.1 | - | 2 | 261 | |||
| AK029491.1 | - | 2 | 239 | |||
| AK122218.1 | - | 2 | 713 | |||
| BC131674.1 | - | 2 | 273 | 40110915 | ||
| BC131675.1 | - | 2 | 273 | 40110921 | ||

Slk
Genomic Map