siRNA Details
Gene Symbol: Tacr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009314.4 | NP_033340.3 | 2 | 835 | ||
GenBank mRNA | AK033443.1 | - | 2 | 518 | ||
BC106959.1 | - | 2 | 484 | 40039032 | ||
X62933.1 | - | 2 | 835 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
