siRNA Details
| Gene Symbol: Slk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164639.1 | NP_001158111.1 | 4 | 606 | ||
| NM_009289.3 | NP_033315.2 | 4 | 606 | |||
| GenBank mRNA | AF039574.1 | - | 4 | 611 | ||
| AF112855.1 | - | 4 | 495 | |||
| AK029491.1 | - | 4 | 473 | |||
| AK122218.1 | - | 4 | 947 | |||
| BC131674.1 | - | 4 | 507 | 40110915 | ||
| BC131675.1 | - | 4 | 507 | 40110921 | ||

Slk
Genomic Map