siRNA Details
| Gene Symbol: Supt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009297.2 | NP_033323.2 | 26 | 3635 | ||
| GenBank mRNA | AK129069.1 | - | 14 | 1958 | ||
| AK158846.1 | - | 8 | 1570 | |||
| AK170533.1 | - | 26 | 3634 | |||
| BC072657.1 | - | 26 | 3555 | 30536338 | ||
| U40375.1 | - | 26 | 3633 | |||

Supt6
Genomic Map