siRNA Details
| Gene Symbol: Sva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009299.2 | NP_033325.2 | 4 | 560 | ||
| GenBank mRNA | CJ055470.1 | - | 4 | 560 | ||
| M94179.1 | - | 4 | 546 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sva| Gene Symbol: Sva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009299.2 | NP_033325.2 | 4 | 560 | ||
| GenBank mRNA | CJ055470.1 | - | 4 | 560 | ||
| M94179.1 | - | 4 | 546 | |||