siRNA Details
| Gene Symbol: Ppp2r5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009358.3 | NP_033384.2 | 14 | 1753 | ||
| GenBank mRNA | AB053464.1 | - | 14 | 1698 | ||
| AF316616.2 | - | 14 | 1610 | |||
| AK154050.1 | - | 14 | 1729 | |||
| AK160587.1 | - | 14 | 1694 | |||
| AK178339.1 | - | 1 | 95 | |||
| AY338234.1 | - | 14 | 1683 | |||
| BC009163.1 | - | 4 | 280 | |||
| BC010716.1 | - | 14 | 1666 | 2655861 | ||
| X81059.1 | - | 3 | 193 | |||

Ppp2r5d
Genomic Map