siRNA Details
| Gene Symbol: Pglyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009402.2 | NP_033428.1 | 2 | 308 | ||
| GenBank mRNA | AF076482.1 | - | 2 | 307 | ||
| AF193843.1 | - | 2 | 296 | |||
| AK008335.1 | - | 2 | 309 | |||
| BC005582.1 | - | 2 | 318 | 3969014 | ||
| CT010165.1 | - | 2 | 272 | |||
| X86374.1 | - | 2 | 307 | |||

Pglyrp1
Genomic Map