siRNA Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009408.2 | NP_033434.2 | 3 | 449 | ||
| GenBank mRNA | AK145538.1 | - | 3 | 459 | ||
| AK160590.1 | - | 3 | 303 | |||
| AK164061.1 | - | 3 | 368 | |||
| BC002095.1 | - | 3 | 330 | |||
| BC048743.1 | - | 3 | 344 | |||
| BC092349.1 | - | 3 | 279 | |||
| D10061.1 | - | 3 | 116 | |||
| L20632.1 | - | 3 | 116 | |||

Top1
Genomic Map