siRNA Details
| Gene Symbol: Tpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009415.2 | NP_033441.2 | 4 | 608 | ||
| GenBank mRNA | AK002270.1 | - | 4 | 470 | ||
| AK010808.1 | - | 4 | 478 | |||
| AK146013.1 | - | 4 | 467 | |||
| AK149768.1 | - | 4 | 472 | |||
| AK150735.1 | - | 4 | 469 | |||
| AK159741.1 | - | 4 | 472 | |||
| AK167437.1 | - | 4 | 473 | |||
| AK168060.1 | - | 4 | 470 | |||
| AK168446.1 | - | 4 | 472 | |||
| AK168756.1 | - | 4 | 471 | |||
| BC004024.1 | - | 4 | 456 | |||
| BC046761.1 | - | 4 | 459 | 5693696 | ||
| BU945752.1 | - | 4 | 614 | |||
| X53333.1 | - | 4 | 470 | |||

Tpi1
Genomic Map