siRNA Details
| Gene Symbol: Tpst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009419.3 | NP_033445.2 | 3 | 413 | ||
| GenBank mRNA | AF049890.1 | - | 2 | 359 | ||
| AK036436.1 | - | 2 | 342 | |||
| AK075672.1 | - | 3 | 413 | |||
| AK149379.1 | - | 3 | 417 | |||
| AK149502.1 | - | 3 | 376 | |||
| AK150082.1 | - | 3 | 376 | |||
| AK163461.1 | - | 3 | 377 | |||
| AK185523.1 | - | 1 | 30 | |||
| BC003721.1 | - | 3 | 404 | 3494892 | ||

Tpst2
Genomic Map