siRNA Details
| Gene Symbol: Tff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009362.2 | NP_033388.1 | 2 | 213 | ||
| GenBank mRNA | AK008625.1 | - | 2 | 210 | ||
| AK008809.1 | - | 2 | 210 | |||
| AK008873.1 | - | 2 | 213 | |||
| AK010179.1 | - | 2 | 210 | |||
| AK131896.1 | - | 2 | 214 | |||
| AK160174.1 | - | 2 | 210 | |||
| BC117054.1 | - | 2 | 210 | 40126373 | ||
| BC117056.1 | - | 2 | 210 | 40126375 | ||
| Z21858.1 | - | 2 | 182 | |||

Tff1
Genomic Map