siRNA Details
| Gene Symbol: Tgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009373.3 | NP_033399.1 | 4 | 608 | ||
| GenBank mRNA | AF076928.1 | - | 3 | 523 | ||
| AF114266.1 | - | 4 | 573 | |||
| AK052912.1 | - | 4 | 583 | |||
| AK054108.1 | - | 4 | 583 | |||
| AK080224.1 | - | 4 | 608 | |||
| AK080593.1 | - | 4 | 583 | |||
| AK086001.1 | - | 4 | 583 | |||
| AK089481.1 | - | 4 | 611 | |||
| AK143712.1 | - | 4 | 644 | |||
| AK151776.1 | - | 4 | 609 | |||
| AK152152.1 | - | 4 | 583 | |||
| AK152627.1 | - | 4 | 609 | |||
| AK159255.1 | - | 4 | 583 | |||
| AK168990.1 | - | 4 | 584 | |||
| AK169356.1 | - | 4 | 611 | |||
| BC016492.1 | - | 4 | 549 | 3256943 | ||
| M55154.1 | - | 4 | 545 | |||

Tgm2
Genomic Map