siRNA Details
| Gene Symbol: Ift88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009376.2 | NP_033402.2 | 5, 6 | 384 | ||
| GenBank mRNA | AK135110.1 | - | 5, 6 | 379 | ||
| AK160633.1 | - | 5, 6 | 393 | |||
| AK168395.1 | - | 4, 5 | 293 | |||
| BC012250.1 | - | 5, 6 | 364 | 4019334 | ||
| L31959.1 | - | 5, 6 | 329 | |||

Ift88
Genomic Map