siRNA Details
| Gene Symbol: Thrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113417.1 | NP_001106888.1 | 5, 6 | 581 | ||
| NM_009380.3 | NP_033406.1 | 2, 3 | 667 | |||
| GenBank mRNA | AB221613.1 | - | 2, 3 | 434 | ||
| AK133662.1 | - | 2, 3 | 164 | |||
| AK158826.1 | - | 5, 6 | 582 | |||
| BC089035.1 | - | 2, 3 | 667 | 30695885 | ||
| BC119552.1 | - | 2, 3 | 604 | 40057420 | ||
| BC119553.1 | - | 2, 3 | 608 | 40057421 | ||
| S62756.1 | - | 4, 5 | 451 | |||

Thrb
Genomic Map