siRNA Details
| Gene Symbol: SHOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000451.3 | NP_000442.1 | 2 | 789 | ||
| NM_006883.2 | NP_006874.1 | 2 | 789 | |||
| GenBank mRNA | U89331.1 | - | 2 | 791 | ||
| Y11535.1 | - | 1 | 189 | |||
| Y11536.1 | - | 1 | 189 | |||

SHOX
Genomic Map