siRNA Details
| Gene Symbol: EPHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005232.4 | NP_005223.4 | 15, 16 | 2594 | ||
| XM_006715880.3 | XP_006715943.1 | 14, 15 | 2222 | |||
| GenBank mRNA | AK290351.1 | - | 15, 16 | 2558 | ||
| AK313263.1 | - | 15, 16 | 2567 | |||
| BC130291.1 | - | 15, 16 | 2578 | 40146322 | ||
| BC144156.1 | - | 13, 14 | 2523 | |||
| M18391.1 | - | 15, 16 | 2624 | |||
| Z27409.1 | - | 11, 12 | 1652 | |||

EPHA1
Genomic Map