siRNA Details
| Gene Symbol: Tfpi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009364.3 | NP_033390.1 | 5 | 1142 | ||
| GenBank mRNA | AK132268.1 | - | 5 | 1164 | ||
| BC021639.1 | - | 5 | 1177 | 4023742 | ||
| D50586.1 | - | 5 | 1151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tfpi2| Gene Symbol: Tfpi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009364.3 | NP_033390.1 | 5 | 1142 | ||
| GenBank mRNA | AK132268.1 | - | 5 | 1164 | ||
| BC021639.1 | - | 5 | 1177 | 4023742 | ||
| D50586.1 | - | 5 | 1151 | |||