siRNA Details
| Gene Symbol: Tnnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009394.2 | NP_033420.1 | 5 | 488 | ||
| GenBank mRNA | AK133548.1 | - | 5 | 484 | ||
| BC024390.1 | - | 5 | 487 | 5026329 | ||
| CB566378.1 | - | 523 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnnc2| Gene Symbol: Tnnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009394.2 | NP_033420.1 | 5 | 488 | ||
| GenBank mRNA | AK133548.1 | - | 5 | 484 | ||
| BC024390.1 | - | 5 | 487 | 5026329 | ||
| CB566378.1 | - | 523 | ||||