siRNA Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009441.2 | NP_033467.2 | 2 | 211 | ||
| GenBank mRNA | AB008516.1 | - | 1 | 88 | ||
| AK053765.1 | - | 2 | 315 | |||
| AK088273.1 | - | 2 | 212 | |||
| AK089974.1 | - | 2 | 154 | |||
| AK166298.1 | - | 2 | 153 | |||
| AK170087.1 | - | 2 | 202 | |||
| AK220495.1 | - | 2 | 253 | |||
| BC019173.1 | - | 2 | 180 | |||
| BC037654.1 | - | 2 | 156 | |||
| BC049277.1 | - | 2 | 193 | |||
| BC057207.1 | - | 2 | 169 | |||
| BC145489.1 | - | 1 | 128 | |||

Ttc3
Genomic Map