siRNA Details
| Gene Symbol: Ube2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169576.1 | NP_001163047.1 | 2 | 515 | ||
| NM_009459.3 | NP_033485.1 | 2 | 515 | |||
| GenBank mRNA | AK131688.1 | - | 2 | 493 | ||
| AK161414.1 | - | 2 | 499 | |||
| BC008517.1 | - | 2 | 321 | 3600233 | ||
| U19854.1 | - | 2 | 244 | |||

Ube2h
Genomic Map