siRNA Details
| Gene Symbol: Unc5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293561.1 | NP_001280490.1 | 2 | 365 | ||
| NM_009472.4 | NP_033498.1 | 2 | 365 | |||
| GenBank mRNA | AK031655.1 | - | 2 | 332 | ||
| AK034558.1 | - | 2 | 332 | |||
| AK035038.1 | - | 2 | 331 | |||
| AK141181.1 | - | 2 | 331 | |||
| AK142411.1 | - | 2 | 331 | |||
| AK142974.1 | - | 2 | 259 | |||
| BC115772.1 | - | 2 | 355 | 40085998 | ||
| U72634.1 | - | 2 | 365 | |||

Unc5c
Genomic Map