siRNA Details
| Gene Symbol: Vmn2r89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009487.1 | NP_033513.1 | 4, 5 | 1598 | ||
| GenBank mRNA | AF011421.1 | - | 4, 5 | 1598 | ||
| BC145855.1 | - | 4, 5 | 1559 | 40131158 | ||
More Information
Set Membership: |
|

Vmn2r89
Genomic Map