siRNA Details
| Gene Symbol: Vmn2r122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009488.2 | NP_033514.2 | 4 | 1443 | ||
| GenBank mRNA | AF011422.1 | - | 4 | 1418 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r122| Gene Symbol: Vmn2r122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009488.2 | NP_033514.2 | 4 | 1443 | ||
| GenBank mRNA | AF011422.1 | - | 4 | 1418 | ||
Set Membership: |
|